Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACAAATATTAAATTCTAGGGGTT[A/T]AAAAAAACTTATGTAAACTTTTAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610148 MIM: 606736 | ||||||||||||||||||||
Literature Links: |
BBS10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BBS10 - Bardet-Biedl syndrome 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OSBPL8 - oxysterol binding protein like 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003712.1 | 3880 | UTR 3 | NP_001003712.1 | |||
NM_001319652.1 | 3880 | UTR 3 | NP_001306581.1 | |||
NM_001319653.1 | 3880 | UTR 3 | NP_001306582.1 | |||
NM_001319655.1 | 3880 | UTR 3 | NP_001306584.1 | |||
NM_020841.4 | 3880 | UTR 3 | NP_065892.1 | |||
XM_005268621.4 | 3880 | UTR 3 | XP_005268678.1 | |||
XM_006719224.2 | 3880 | UTR 3 | XP_006719287.1 | |||
XM_011537855.2 | 3880 | UTR 3 | XP_011536157.1 | |||
XM_011537856.2 | 3880 | UTR 3 | XP_011536158.1 | |||
XM_011537857.2 | 3880 | UTR 3 | XP_011536159.1 | |||
XM_017018768.1 | 3880 | UTR 3 | XP_016874257.1 | |||
XM_017018769.1 | 3880 | UTR 3 | XP_016874258.1 |