Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAAAGAGACGAGATGAGACGAGA[A/T]GAGAAGAGAAGAAGGAAGGAAGTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610776 | ||||||||||||||||||||
Literature Links: |
DRAM1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DRAM1 - DNA damage regulated autophagy modulator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018370.2 | Intron | NP_060840.2 | ||||
XM_005269004.2 | Intron | XP_005269061.1 | ||||
XM_005269005.2 | Intron | XP_005269062.1 | ||||
XM_017019578.1 | Intron | XP_016875067.1 |