Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAAGGACCTTATGATCTAGTAGAA[G/T]TTTATTCAGGGCTTCAGATTTAGAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
29 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604903 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TAF1A PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TAF1A - TATA-box binding protein associated factor, RNA polymerase I subunit A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201536.1 | Intron | NP_001188465.1 | ||||
NM_005681.3 | Intron | NP_005672.1 | ||||
NM_139352.2 | Intron | NP_647603.1 | ||||
XM_005273343.3 | Intron | XP_005273400.1 | ||||
XM_006711612.1 | Intron | XP_006711675.1 | ||||
XM_006711613.3 | Intron | XP_006711676.1 | ||||
XM_017002758.1 | Intron | XP_016858247.1 | ||||
XM_017002759.1 | Intron | XP_016858248.1 | ||||
XM_017002760.1 | Intron | XP_016858249.1 |