Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
38 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612375 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AIDA PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
AIDA - axin interactor, dorsalization associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022831.2 | 282 | Intron | NP_073742.2 |
BROX - BRO1 domain and CAAX motif containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288579.1 | 282 | Intron | NP_001275508.1 | |||
NM_001288580.1 | 282 | Intron | NP_001275509.1 | |||
NM_001288581.1 | 282 | Intron | NP_001275510.1 | |||
NM_144695.3 | 282 | Intron | NP_653296.2 | |||
XM_005273065.2 | 282 | Intron | XP_005273122.2 | |||
XM_005273069.4 | 282 | Intron | XP_005273126.1 | |||
XM_006711173.3 | 282 | Intron | XP_006711236.1 | |||
XM_011509212.2 | 282 | UTR 5 | XP_011507514.1 | |||
XM_011509213.2 | 282 | UTR 5 | XP_011507515.1 | |||
XM_011509214.2 | 282 | Intron | XP_011507516.1 | |||
XM_017000374.1 | 282 | Intron | XP_016855863.1 | |||
XM_017000375.1 | 282 | UTR 5 | XP_016855864.1 | |||
XM_017000376.1 | 282 | Intron | XP_016855865.1 |