Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTCAAAAAAAAAAAAAAAAAAA[A/G]AGGGGCGATGTTTATGGAGAAAGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 606225 | ||||||||||||||||||||
Literature Links: |
NOL9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NOL9 - nucleolar protein 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAS1R1 - taste 1 receptor member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138697.3 | Intron | NP_619642.2 | ||||
NM_177540.2 | Intron | NP_803884.1 | ||||
XM_011542203.2 | Intron | XP_011540505.1 | ||||
XM_011542206.2 | Intron | XP_011540508.1 | ||||
XM_017002402.1 | Intron | XP_016857891.1 | ||||
XM_017002403.1 | Intron | XP_016857892.1 |