Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGAGGCCTGGCTTCCAGTATAGC[C/T]GGAGCCCAGGATCCAGAGTGTGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
30 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610371 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC2A7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC2A7 - solute carrier family 2 member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207420.2 | 1548 | Intron | NP_997303.2 | |||
XM_011540824.2 | 1548 | Silent Mutation | CCA,CCG | P,P 503 | XP_011539126.1 | |
XM_011540825.2 | 1548 | Intron | XP_011539127.1 |