Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTATATATATATTTTTTTTTTTGG[A/G]AGGATTTGTGGTAAGTTACAGACAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 601882 MIM: 601791 | ||||||||||||||||||||
Literature Links: |
DFFA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DFFA - DNA fragmentation factor subunit alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PEX14 - peroxisomal biogenesis factor 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004565.2 | Intron | NP_004556.1 | ||||
XM_005263470.4 | Intron | XP_005263527.1 | ||||
XM_011541577.2 | Intron | XP_011539879.1 | ||||
XM_011541578.2 | Intron | XP_011539880.1 | ||||
XM_011541579.2 | Intron | XP_011539881.1 | ||||
XM_011541580.1 | Intron | XP_011539882.1 |