Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGACCAAGAGCGCACTCCAGCCC[C/T]GAGCCCAGAGCCTGCAAAGCCCAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
20 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607093 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf167 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1orf167 - chromosome 1 open reading frame 167 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010881.1 | 4095 | Silent Mutation | CCC,CCT | P,P 1266 | NP_001010881.1 | |
XM_011541269.2 | 4095 | Silent Mutation | CCC,CCT | P,P 1311 | XP_011539571.2 | |
XM_011541271.2 | 4095 | Silent Mutation | CCC,CCT | P,P 1293 | XP_011539573.2 | |
XM_011541272.2 | 4095 | Silent Mutation | CCC,CCT | P,P 1311 | XP_011539574.1 | |
XM_011541274.2 | 4095 | Silent Mutation | CCC,CCT | P,P 1266 | XP_011539576.2 | |
XM_011541275.2 | 4095 | Silent Mutation | CCC,CCT | P,P 1266 | XP_011539577.2 | |
XM_011541276.2 | 4095 | Silent Mutation | CCC,CCT | P,P 1311 | XP_011539578.1 | |
XM_011541277.2 | 4095 | Silent Mutation | CCC,CCT | P,P 1311 | XP_011539579.1 | |
XM_011541278.2 | 4095 | Silent Mutation | CCC,CCT | P,P 1311 | XP_011539580.1 | |
XM_011541280.2 | 4095 | Silent Mutation | CCC,CCT | P,P 738 | XP_011539582.2 | |
XM_011541281.2 | 4095 | Silent Mutation | CCC,CCT | P,P 738 | XP_011539583.2 | |
XM_017001062.1 | 4095 | Silent Mutation | CCC,CCT | P,P 1311 | XP_016856551.1 | |
XM_017001063.1 | 4095 | Silent Mutation | CCC,CCT | P,P 1311 | XP_016856552.1 | |
XM_017001064.1 | 4095 | Silent Mutation | CCC,CCT | P,P 1311 | XP_016856553.1 |
LOC102724659 - uncharacterized LOC102724659 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MTHFR - methylenetetrahydrofolate reductase (NAD(P)H) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005957.4 | 4095 | UTR 3 | NP_005948.3 | |||
XM_005263458.3 | 4095 | Intron | XP_005263515.1 | |||
XM_005263460.4 | 4095 | Intron | XP_005263517.1 | |||
XM_005263462.4 | 4095 | Intron | XP_005263519.1 | |||
XM_005263463.3 | 4095 | Intron | XP_005263520.1 | |||
XM_011541495.2 | 4095 | Intron | XP_011539797.1 | |||
XM_011541496.2 | 4095 | Intron | XP_011539798.1 | |||
XM_017001328.1 | 4095 | Intron | XP_016856817.1 |