Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACTGTCTCCAGAAGCTTCCCTCC[C/T]CACGACAGATAATGAGCAGCAAGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615296 MIM: 605508 MIM: 605507 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL1F10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL1F10 - interleukin 1 family member 10 (theta) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IL36B - interleukin 36, beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IL36RN - interleukin 36 receptor antagonist | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012275.2 | Intron | NP_036407.1 | ||||
NM_173170.1 | Intron | NP_775262.1 |