Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGGTGGCAAAATAAATTGGTAGC[A/G]GAGCTAGGTCTAGGACTCAGCCTTT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 615296 MIM: 605507 | |||||||||||||||||||||||
Literature Links: |
IL1F10 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
IL1F10 - interleukin 1 family member 10 (theta) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032556.5 | Intron | NP_115945.4 | ||||
NM_173161.2 | Intron | NP_775184.1 |
IL36RN - interleukin 36 receptor antagonist | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |