Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTGAGTCAGGAGAAGTAGGAGAT[A/G]TTGTCAATGAGTCTCAGGTTTTGAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615296 MIM: 605507 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL1F10 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IL1F10 - interleukin 1 family member 10 (theta) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032556.5 | Intron | NP_115945.4 | ||||
NM_173161.2 | Intron | NP_775184.1 |
IL36RN - interleukin 36 receptor antagonist | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |