Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGCTGTGCCTTAAGCAGAAGAAG[C/T]ATACATCTGACCCTCAAAGGGACAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607306 | ||||||||||||||||||||
Literature Links: |
SRD5A2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SRD5A2 - steroid 5 alpha-reductase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000348.3 | 4341 | Intron | NP_000339.2 | |||
XM_011533069.2 | 4341 | UTR 3 | XP_011531371.1 | |||
XM_011533072.2 | 4341 | UTR 3 | XP_011531374.1 |