Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGAATGCCCTTTCATGTTTTTGA[C/G]GTTTGCCTTTAACAACTTTTACTCC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
FRMD4B PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
FRMD4B - FERM domain containing 4B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015123.2 | 2907 | UTR 3 | NP_055938.1 | |||
XM_005264722.1 | 2907 | UTR 3 | XP_005264779.1 | |||
XM_005264723.1 | 2907 | UTR 3 | XP_005264780.1 | |||
XM_017005989.1 | 2907 | UTR 3 | XP_016861478.1 | |||
XM_017005990.1 | 2907 | UTR 3 | XP_016861479.1 | |||
XM_017005991.1 | 2907 | UTR 3 | XP_016861480.1 | |||
XM_017005992.1 | 2907 | UTR 3 | XP_016861481.1 | |||
XM_017005993.1 | 2907 | UTR 3 | XP_016861482.1 | |||
XM_017005994.1 | 2907 | UTR 3 | XP_016861483.1 | |||
XM_017005995.1 | 2907 | UTR 3 | XP_016861484.1 | |||
XM_017005996.1 | 2907 | UTR 3 | XP_016861485.1 | |||
XM_017005997.1 | 2907 | UTR 3 | XP_016861486.1 |