Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
MIR4273 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR4273 - microRNA 4273 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF717 - zinc finger protein 717 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128223.2 | Intron | NP_001121695.1 | ||||
NM_001290208.1 | Intron | NP_001277137.1 | ||||
NM_001290209.1 | Intron | NP_001277138.1 | ||||
NM_001290210.1 | Intron | NP_001277139.1 | ||||
NM_001324026.1 | Intron | NP_001310955.1 | ||||
NM_001324027.1 | Intron | NP_001310956.1 | ||||
NM_001324028.1 | Intron | NP_001310957.1 |