Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATATAATATAGTGTATATATAGTA[G/T]GGTATATATATACACATATATGATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608260 | ||||||||||||||||||||
Literature Links: |
KCNH8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KCNH8 - potassium voltage-gated channel subfamily H member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144633.2 | Intron | NP_653234.2 | ||||
XM_011533347.2 | Intron | XP_011531649.1 | ||||
XM_017005698.1 | Intron | XP_016861187.1 | ||||
XM_017005699.1 | Intron | XP_016861188.1 | ||||
XM_017005700.1 | Intron | XP_016861189.1 | ||||
XM_017005701.1 | Intron | XP_016861190.1 | ||||
XM_017005702.1 | Intron | XP_016861191.1 |