Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATGTCAAATACACTAAATATATA[C/T]ATGTGTACATACACAGGTACCTGGA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 605458 | |||||||||||||||||||||||
Literature Links: |
CHDH PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CHDH - choline dehydrogenase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IL17RB - interleukin 17 receptor B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018725.3 | Intron | NP_061195.2 | ||||
XM_005265310.4 | Intron | XP_005265367.1 | ||||
XM_005265311.4 | Intron | XP_005265368.1 | ||||
XM_005265312.4 | Intron | XP_005265369.1 | ||||
XM_011533941.2 | Intron | XP_011532243.1 | ||||
XM_017006804.1 | Intron | XP_016862293.1 | ||||
XM_017006805.1 | Intron | XP_016862294.1 | ||||
XM_017006806.1 | Intron | XP_016862295.1 | ||||
XM_017006807.1 | Intron | XP_016862296.1 |