Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATCCATAGCAACACTGTGTGTAA[C/T]AGCAAAGACAAGAAACAAACACAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613974 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDX60 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DDX60 - DEXD/H-box helicase 60 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017631.5 | Intron | NP_060101.3 | ||||
XM_011532103.2 | Intron | XP_011530405.1 | ||||
XM_011532104.2 | Intron | XP_011530406.1 | ||||
XM_017008383.1 | Intron | XP_016863872.1 | ||||
XM_017008384.1 | Intron | XP_016863873.1 |