Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGACGGGTTGAAAGTATTTAGATGC[G/T]TTCTGTTCCTGAGGTCGATCATATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
STOX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
STOX2 - storkhead box 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020225.2 | Intron | NP_064610.1 | ||||
XM_011532129.1 | Intron | XP_011530431.1 | ||||
XM_011532130.1 | Intron | XP_011530432.1 | ||||
XM_011532131.1 | Intron | XP_011530433.1 | ||||
XM_017008466.1 | Intron | XP_016863955.1 | ||||
XM_017008467.1 | Intron | XP_016863956.1 |