Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612404 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RASL11B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RASL11B - RAS like family 11 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCFD2 - sec1 family domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152540.3 | 2469 | UTR 3 | NP_689753.2 | |||
XM_011534375.2 | 2469 | Intron | XP_011532677.1 | |||
XM_011534376.2 | 2469 | Intron | XP_011532678.1 | |||
XM_011534378.2 | 2469 | Intron | XP_011532680.1 | |||
XM_017007786.1 | 2469 | Intron | XP_016863275.1 | |||
XM_017007787.1 | 2469 | Intron | XP_016863276.1 |
Set Membership: |
HapMap |