Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACTGTGCTGAATTGCAAGGAGTT[C/T]CCCCCACACGTGTTATTTCCACAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612849 | ||||||||||||||||||||
Literature Links: |
USP46 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
USP46 - ubiquitin specific peptidase 46 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134223.1 | 6774 | UTR 3 | NP_001127695.1 | |||
NM_001286767.1 | 6774 | UTR 3 | NP_001273696.1 | |||
NM_001286768.1 | 6774 | UTR 3 | NP_001273697.1 | |||
NM_022832.3 | 6774 | UTR 3 | NP_073743.2 | |||
XM_017008557.1 | 6774 | UTR 3 | XP_016864046.1 |