Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATATAACATGTCAGTCCAGCCAA[G/T]CGCGGTGGCTCCCACCTGTAATCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182309 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC34A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC34A1 - solute carrier family 34 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167579.1 | Intron | NP_001161051.1 | ||||
NM_003052.4 | Intron | NP_003043.3 | ||||
XM_005265975.1 | Intron | XP_005266032.1 | ||||
XM_017009773.1 | Intron | XP_016865262.1 | ||||
XM_017009774.1 | Intron | XP_016865263.1 | ||||
XM_017009775.1 | Intron | XP_016865264.1 |