Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGCTTGAACTCCTGGGCTCAAGC[A/G]TTGTGCCTGCCTCAGCCTCCCAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114217 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CANX PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CANX - calnexin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024649.1 | Intron | NP_001019820.1 | ||||
NM_001746.3 | Intron | NP_001737.1 | ||||
XM_011534664.1 | Intron | XP_011532966.1 | ||||
XM_011534665.2 | Intron | XP_011532967.1 |
CBY3 - chibby family member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |