Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACAGCCAGGGTTTCCAGACTGCT[C/T]TCCTGCTCTCACAGAACTCTGTCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602896 | ||||||||||||||||||||
Literature Links: |
MAPK9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAPK9 - mitogen-activated protein kinase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135044.1 | 4016 | Intron | NP_001128516.1 | |||
NM_001308244.1 | 4016 | Intron | NP_001295173.1 | |||
NM_002752.4 | 4016 | UTR 3 | NP_002743.3 | |||
NM_139068.2 | 4016 | UTR 3 | NP_620707.1 | |||
NM_139069.2 | 4016 | UTR 3 | NP_620708.1 | |||
NM_139070.2 | 4016 | UTR 3 | NP_620709.1 | |||
XM_005265940.4 | 4016 | UTR 3 | XP_005265997.1 | |||
XM_006714891.2 | 4016 | UTR 3 | XP_006714954.1 | |||
XM_017009638.1 | 4016 | UTR 3 | XP_016865127.1 | |||
XM_017009639.1 | 4016 | UTR 3 | XP_016865128.1 | |||
XM_017009640.1 | 4016 | UTR 3 | XP_016865129.1 | |||
XM_017009641.1 | 4016 | UTR 3 | XP_016865130.1 | |||
XM_017009642.1 | 4016 | Intron | XP_016865131.1 | |||
XM_017009643.1 | 4016 | Intron | XP_016865132.1 |