Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGTTTAGGAACAGAAGGAAGGCA[A/G]TTTTTGCGTGACTCAGTTCACAAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612722 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ELP3 - elongator acetyltransferase complex subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284220.1 | Intron | NP_001271149.1 | ||||
NM_001284222.1 | Intron | NP_001271151.1 | ||||
NM_001284224.1 | Intron | NP_001271153.1 | ||||
NM_001284225.1 | Intron | NP_001271154.1 | ||||
NM_001284226.1 | Intron | NP_001271155.1 | ||||
NM_018091.5 | Intron | NP_060561.3 | ||||
XM_006716354.2 | Intron | XP_006716417.1 | ||||
XM_017013604.1 | Intron | XP_016869093.1 |
NUGGC - nuclear GTPase, germinal center associated | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |