Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTACAGGTGGCCAACAGGTACAT[G/T]AAAAAATATTCGACATCGTGAATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600712 MIM: 610404 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HNRNPK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HNRNPK - heterogeneous nuclear ribonucleoprotein K | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RMI1 - RecQ mediated genome instability 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024945.2 | Intron | NP_079221.2 | ||||
XM_005252211.2 | Intron | XP_005252268.1 | ||||
XM_005252213.2 | Intron | XP_005252270.1 | ||||
XM_011519034.2 | Intron | XP_011517336.1 | ||||
XM_017015140.1 | Intron | XP_016870629.1 |