Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGTGCAGAAAGGCACCTACGGC[C/T]CTGGAAGCCCAGTTGCGGAAGGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616748 MIM: 611255 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENTPD8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ENTPD8 - ectonucleoside triphosphate diphosphohydrolase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033113.1 | 1558 | UTR 3 | NP_001028285.1 | |||
NM_198585.2 | 1558 | UTR 3 | NP_940987.2 | |||
XM_011518665.2 | 1558 | UTR 3 | XP_011516967.1 | |||
XM_011518666.2 | 1558 | UTR 3 | XP_011516968.1 | |||
XM_011518668.1 | 1558 | UTR 3 | XP_011516970.1 |
NOXA1 - NADPH oxidase activator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256067.1 | 1558 | Intron | NP_001242996.1 | |||
NM_001256068.1 | 1558 | Intron | NP_001242997.1 | |||
NM_006647.1 | 1558 | Intron | NP_006638.1 | |||
XM_011518158.2 | 1558 | Intron | XP_011516460.1 | |||
XM_011518159.1 | 1558 | Intron | XP_011516461.1 | |||
XM_011518160.1 | 1558 | Intron | XP_011516462.1 | |||
XM_017014220.1 | 1558 | Intron | XP_016869709.1 |
Set Membership: |
HapMap |