Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGCATCTGCTCAGGGGCCAGCAT[A/G]TAGGTGACGGCCTTGGGGAGGGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609491 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DKFZP434A062 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DKFZP434A062 - uncharacterized LOC26102 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GPSM1 - G-protein signaling modulator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145638.2 | Intron | NP_001139110.2 | ||||
NM_001145639.1 | Intron | NP_001139111.1 | ||||
NM_001200003.1 | Intron | NP_001186932.1 | ||||
NM_015597.5 | Intron | NP_056412.5 | ||||
XM_011518498.2 | Intron | XP_011516800.1 | ||||
XM_017014598.1 | Intron | XP_016870087.1 | ||||
XM_017014599.1 | Intron | XP_016870088.1 |