Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGGGTGAACCAGTTCCAAATCTC[A/C]TGCCCATTTGCAGTGGTATCACCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606655 | ||||||||||||||||||||
Literature Links: |
RXFP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RXFP2 - relaxin/insulin like family peptide receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166058.1 | Intron | NP_001159530.1 | ||||
NM_130806.3 | Intron | NP_570718.1 | ||||
XM_017020389.1 | Intron | XP_016875878.1 |