Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACATGTATTTTCCTCCTCAAGTTC[A/G]GGCATCAGAGTATGATTAGTGACTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611449 | ||||||||||||||||||||
Literature Links: |
EEF1AKMT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EEF1AKMT1 - eukaryotic translation elongation factor 1 alpha lysine methyltransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
XPO4 - exportin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022459.4 | 4473 | UTR 3 | NP_071904.4 | |||
XM_005266502.1 | 4473 | UTR 3 | XP_005266559.1 | |||
XM_005266503.3 | 4473 | UTR 3 | XP_005266560.1 | |||
XM_005266504.3 | 4473 | UTR 3 | XP_005266561.1 | |||
XM_011535197.1 | 4473 | UTR 3 | XP_011533499.1 | |||
XM_011535198.1 | 4473 | UTR 3 | XP_011533500.1 | |||
XM_011535199.1 | 4473 | UTR 3 | XP_011533501.1 | |||
XM_011535200.2 | 4473 | UTR 3 | XP_011533502.1 | |||
XM_017020704.1 | 4473 | UTR 3 | XP_016876193.1 | |||
XM_017020705.1 | 4473 | UTR 3 | XP_016876194.1 | |||
XM_017020706.1 | 4473 | UTR 3 | XP_016876195.1 |