Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACAGAAATAATTTCCTACCTACA[A/C]TGACAGTATATTTAAACTAAAAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605718 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MED4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MED4 - mediator complex subunit 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270629.1 | 1468 | UTR 3 | NP_001257558.1 | |||
NM_014166.3 | 1468 | UTR 3 | NP_054885.1 | |||
XM_017020549.1 | 1468 | UTR 3 | XP_016876038.1 |
MED4-AS1 - MED4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |