Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGCATATCACAACGTACGTGTCCT[C/T]CTCATCTAGCCCAGGCTCAAGTCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604044 MIM: 616731 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101929657 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101929657 - uncharacterized LOC101929657 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NEK3 - NIMA related kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146099.1 | 1754 | Missense Mutation | AAG,GAG | K,E 460 | NP_001139571.1 | |
NM_002498.2 | 1754 | Missense Mutation | AAG,GAG | K,E 477 | NP_002489.1 | |
NM_152720.2 | 1754 | Missense Mutation | AAG,GAG | K,E 477 | NP_689933.1 |
NEK5 - NIMA related kinase 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |