Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGTTTGGTGAGTTTTGACAAATC[A/G]GTCACTACCACAATTAAAATACAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606097 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA1468 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIAA1468 - KIAA1468 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020854.3 | Intron | NP_065905.2 | ||||
XM_005266730.2 | Intron | XP_005266787.1 | ||||
XM_011526110.1 | Intron | XP_011524412.1 | ||||
XM_011526111.1 | Intron | XP_011524413.1 | ||||
XM_011526112.1 | Intron | XP_011524414.1 | ||||
XM_011526114.1 | Intron | XP_011524416.1 | ||||
XM_011526117.2 | Intron | XP_011524419.1 | ||||
XM_017025882.1 | Intron | XP_016881371.1 | ||||
XM_017025883.1 | Intron | XP_016881372.1 |
PIGN - phosphatidylinositol glycan anchor biosynthesis class N | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |