Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCCGGGAAGGGCCGGGGCCGGCG[A/G]CATCCTGGCTCCTCCGCCTTCCGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603499 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TNFRSF11A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TNFRSF11A - TNF receptor superfamily member 11a | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270949.1 | Intron | NP_001257878.1 | ||||
NM_001270950.1 | Intron | NP_001257879.1 | ||||
NM_001270951.1 | Intron | NP_001257880.1 | ||||
NM_001278268.1 | Intron | NP_001265197.1 | ||||
NM_003839.3 | Intron | NP_003830.1 | ||||
XM_011526244.2 | Intron | XP_011524546.1 | ||||
XM_011526245.2 | Intron | XP_011524547.1 | ||||
XM_017026064.1 | Intron | XP_016881553.1 | ||||
XM_017026065.1 | Intron | XP_016881554.1 | ||||
XM_017026066.1 | Intron | XP_016881555.1 |