Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGAAACTCCTCACTCCCATACAT[A/T]CAAGTCCCCCATACACACAAAGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603571 MIM: 606592 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
VNN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
VNN2 - vanin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242350.2 | 1967 | UTR 3 | NP_001229279.1 | |||
NM_004665.4 | 1967 | UTR 3 | NP_004656.2 | |||
NM_078488.2 | 1967 | UTR 3 | NP_511043.1 | |||
XM_006715593.3 | 1967 | UTR 3 | XP_006715656.1 | |||
XM_011536231.1 | 1967 | Intron | XP_011534533.1 | |||
XM_017011407.1 | 1967 | UTR 3 | XP_016866896.1 | |||
XM_017011408.1 | 1967 | UTR 3 | XP_016866897.1 | |||
XM_017011409.1 | 1967 | UTR 3 | XP_016866898.1 |
VNN3 - vanin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |