Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCTCTTCTATGAGTAGGAAATTC[A/G]GAGGTTAAGTGACTTTCCCAAGGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603571 | ||||||||||||||||||||
Literature Links: |
VNN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
VNN2 - vanin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242350.2 | Intron | NP_001229279.1 | ||||
NM_004665.4 | Intron | NP_004656.2 | ||||
NM_078488.2 | Intron | NP_511043.1 | ||||
XM_006715593.3 | Intron | XP_006715656.1 | ||||
XM_011536231.1 | Intron | XP_011534533.1 | ||||
XM_017011407.1 | Intron | XP_016866896.1 | ||||
XM_017011408.1 | Intron | XP_016866897.1 | ||||
XM_017011409.1 | Intron | XP_016866898.1 |