Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTATCATTAATCAACTCTTCATT[G/T]GAAAAACACACACGAGGGAATTACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604645 | ||||||||||||||||||||
Literature Links: |
LOC101928326 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC101928326 - uncharacterized LOC101928326 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDE7B - phosphodiesterase 7B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018945.3 | Intron | NP_061818.1 | ||||
XM_005266931.3 | Intron | XP_005266988.1 | ||||
XM_011535755.2 | Intron | XP_011534057.1 | ||||
XM_017010741.1 | Intron | XP_016866230.1 |