Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGATATTATATGGGAAAGTGTAAG[A/G]AAAGCACTGAACCACCTGAAGGTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606439 MIM: 607203 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATL1 - atlastin GTPase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SAV1 - salvador family WW domain containing protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021818.3 | Intron | NP_068590.1 | ||||
XM_011537057.2 | Intron | XP_011535359.1 | ||||
XM_017021568.1 | Intron | XP_016877057.1 |