Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAATAAGCATCATATGATATGATC[A/T]GTCCCCACAGGCCTGGGAGGTAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601663 MIM: 608442 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ESR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ESR2 - estrogen receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040275.1 | 4058 | Intron | NP_001035365.1 | |||
NM_001214902.1 | 4058 | Intron | NP_001201831.1 | |||
NM_001271876.1 | 4058 | Intron | NP_001258805.1 | |||
NM_001271877.1 | 4058 | Intron | NP_001258806.1 | |||
NM_001291712.1 | 4058 | Intron | NP_001278641.1 | |||
NM_001291723.1 | 4058 | Intron | NP_001278652.1 | |||
NM_001437.2 | 4058 | Intron | NP_001428.1 | |||
XM_017021079.1 | 4058 | UTR 3 | XP_016876568.1 | |||
XM_017021080.1 | 4058 | UTR 3 | XP_016876569.1 | |||
XM_017021081.1 | 4058 | UTR 3 | XP_016876570.1 | |||
XM_017021082.1 | 4058 | UTR 3 | XP_016876571.1 | |||
XM_017021083.1 | 4058 | UTR 3 | XP_016876572.1 | |||
XM_017021084.1 | 4058 | Intron | XP_016876573.1 |
SYNE2 - spectrin repeat containing nuclear envelope protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |