Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCAATCCTCCATCTGCCTCCAGG[G/T]AGAAACTCTGCCATGGGAAGACCGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616761 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SUSD6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SUSD6 - sushi domain containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014734.3 | Intron | NP_055549.1 | ||||
XM_011537412.1 | Intron | XP_011535714.1 | ||||
XM_011537413.1 | Intron | XP_011535715.1 | ||||
XM_017021838.1 | Intron | XP_016877327.1 | ||||
XM_017021839.1 | Intron | XP_016877328.1 |