Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGACATTCCTGTTATGTTCTCCA[C/T]GTTCTCCTTCACTGAAGATTATAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606454 MIM: 604395 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF2B2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EIF2B2 - eukaryotic translation initiation factor 2B subunit beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MLH3 - mutL homolog 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040108.1 | Intron | NP_001035197.1 | ||||
NM_014381.2 | Intron | NP_055196.2 | ||||
XM_005267532.4 | Intron | XP_005267589.1 | ||||
XM_005267533.4 | Intron | XP_005267590.1 | ||||
XM_005267534.2 | Intron | XP_005267591.1 | ||||
XM_006720116.3 | Intron | XP_006720179.1 | ||||
XM_011536646.2 | Intron | XP_011534948.1 | ||||
XM_011536648.2 | Intron | XP_011534950.1 | ||||
XM_017021219.1 | Intron | XP_016876708.1 |