Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAATGGCTGATGGAGGCGCTGATT[C/G]GGCAGCGCAAAGACTTCCAGAAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602699 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NFATC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NFATC4 - nuclear factor of activated T-cells 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136022.2 | Intron | NP_001129494.1 | ||||
NM_001198965.1 | Intron | NP_001185894.1 | ||||
NM_001198966.2 | Intron | NP_001185895.1 | ||||
NM_001198967.2 | Intron | NP_001185896.1 | ||||
NM_001288802.1 | Intron | NP_001275731.1 | ||||
NM_001320043.1 | Intron | NP_001306972.1 | ||||
NM_004554.4 | Intron | NP_004545.2 | ||||
XM_011536797.2 | Intron | XP_011535099.1 | ||||
XM_011536798.1 | Intron | XP_011535100.1 | ||||
XM_011536799.2 | Intron | XP_011535101.1 |