Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGCCGAGGGCAGCCTCTTCCTCG[C/T]GGAGCCGGTCAGCCAGCAGTCTCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600025 MIM: 600675 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLC1 - kinesin light chain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130107.1 | 1285 | Intron | NP_001123579.1 | |||
NM_005552.4 | 1285 | Intron | NP_005543.2 | |||
NM_182923.3 | 1285 | Intron | NP_891553.2 |
XRCC3 - X-ray repair cross complementing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100118.1 | 1285 | Missense Mutation | NP_001093588.1 | |||
NM_001100119.1 | 1285 | Missense Mutation | NP_001093589.1 | |||
NM_005432.3 | 1285 | Missense Mutation | NP_005423.1 | |||
XM_005268046.2 | 1285 | Missense Mutation | XP_005268103.1 | |||
XM_011537138.2 | 1285 | Missense Mutation | XP_011535440.1 |