Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGCCTATGAGGGAGAGGATCAGG[A/G]TGGGGTGATGGAGGATCCGTGGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606455 MIM: 600675 MIM: 613504 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP1R13B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPP1R13B - protein phosphatase 1 regulatory subunit 13B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
XRCC3 - X-ray repair cross complementing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZFYVE21 - zinc finger FYVE-type containing 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198953.1 | Intron | NP_001185882.1 | ||||
NM_024071.3 | Intron | NP_076976.1 |