Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGAAGTCCTTAACATTTCCCTAC[A/G]TGAATCGGATTGTTCTGAGGGCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164730 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AKT1 - AKT serine/threonine kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014431.1 | 2051 | UTR 3 | NP_001014431.1 | |||
NM_001014432.1 | 2051 | UTR 3 | NP_001014432.1 | |||
NM_005163.2 | 2051 | UTR 3 | NP_005154.2 | |||
XM_005267401.1 | 2051 | UTR 3 | XP_005267458.1 | |||
XM_017021075.1 | 2051 | Intron | XP_016876564.1 | |||
XM_017021076.1 | 2051 | Intron | XP_016876565.1 | |||
XM_017021077.1 | 2051 | Intron | XP_016876566.1 | |||
XM_017021078.1 | 2051 | Intron | XP_016876567.1 |