Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACAGCGGTGCAGGACAGTCAGGC[A/T]GTACAGGACAGTCAGGCTGTACAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604902 MIM: 610423 | ||||||||||||||||||||
Literature Links: |
BRF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BRF1 - BRF1, RNA polymerase III transcription initiation factor 90 kDa subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242786.1 | Intron | NP_001229715.1 | ||||
NM_001242787.1 | Intron | NP_001229716.1 | ||||
NM_001242788.1 | Intron | NP_001229717.1 | ||||
NM_001242789.1 | Intron | NP_001229718.1 | ||||
NM_001242790.1 | Intron | NP_001229719.1 | ||||
NM_001519.3 | Intron | NP_001510.2 | ||||
NM_145685.2 | Intron | NP_663718.1 | ||||
XM_005267561.3 | Intron | XP_005267618.1 | ||||
XM_005267563.3 | Intron | XP_005267620.1 | ||||
XM_006720123.2 | Intron | XP_006720186.1 | ||||
XM_011536672.2 | Intron | XP_011534974.1 | ||||
XM_011536673.1 | Intron | XP_011534975.1 | ||||
XM_011536674.1 | Intron | XP_011534976.1 |
PACS2 - phosphofurin acidic cluster sorting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100913.2 | Intron | NP_001094383.2 | ||||
NM_001243127.2 | Intron | NP_001230056.1 | ||||
NM_015197.3 | Intron | NP_056012.2 | ||||
XM_006720090.1 | Intron | XP_006720153.1 | ||||
XM_006720091.1 | Intron | XP_006720154.1 | ||||
XM_006720092.2 | Intron | XP_006720155.1 | ||||
XM_017021105.1 | Intron | XP_016876594.1 | ||||
XM_017021106.1 | Intron | XP_016876595.1 | ||||
XM_017021107.1 | Intron | XP_016876596.1 | ||||
XM_017021108.1 | Intron | XP_016876597.1 | ||||
XM_017021109.1 | Intron | XP_016876598.1 | ||||
XM_017021110.1 | Intron | XP_016876599.1 | ||||
XM_017021111.1 | Intron | XP_016876600.1 | ||||
XM_017021112.1 | Intron | XP_016876601.1 |