Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGAAAATCCATAATACAGAGATG[C/T]ATTATTTTCTTACTCCTTTATGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182279 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNRPN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SNRPN - small nuclear ribonucleoprotein polypeptide N | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003097.4 | Intron | NP_003088.1 | ||||
NM_022805.3 | Intron | NP_073716.1 | ||||
NM_022806.3 | Intron | NP_073717.1 | ||||
NM_022807.3 | Intron | NP_073718.1 | ||||
NM_022808.3 | Intron | NP_073719.1 |