Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACAGGGCAAGAGCGCCTTGTGTTA[G/T]CTTCTTCCCTAATTGGGGATGTTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137142 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GABRA5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GABRA5 - gamma-aminobutyric acid type A receptor alpha5 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000810.3 | Intron | NP_000801.1 | ||||
NM_001165037.1 | Intron | NP_001158509.1 | ||||
XM_005268258.2 | Intron | XP_005268315.1 | ||||
XM_006720459.2 | Intron | XP_006720522.1 | ||||
XM_017022055.1 | Intron | XP_016877544.1 | ||||
XM_017022056.1 | Intron | XP_016877545.1 |