Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCTTGTGTTTCCATGTGAATTTT[A/G]GAATCGGCCAATTTGTATAAGAAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603137 MIM: 613713 | ||||||||||||||||||||
Literature Links: |
CUL4A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CUL4A - cullin 4A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008895.2 | Intron | NP_001008895.1 | ||||
NM_001278513.1 | Intron | NP_001265442.1 | ||||
NM_001278514.1 | Intron | NP_001265443.1 | ||||
NM_003589.2 | Intron | NP_003580.1 | ||||
XM_011537523.2 | Intron | XP_011535825.1 |
PCID2 - PCI domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |