Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCCTCCACCTCCCACTGCTCTTC[C/T]TTCAGCCTCTGGCAGCAGTGCAGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610302 | ||||||||||||||||||||
Literature Links: |
EDEM2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EDEM2 - ER degradation enhancing alpha-mannosidase like protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145025.1 | 1465 | Silent Mutation | AAA,AAG | K,K 455 | NP_001138497.1 | |
NM_018217.2 | 1465 | Silent Mutation | AAA,AAG | K,K 492 | NP_060687.2 |