Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCACCATGGAGCCAGGCGGGGCC[C/T]GTCTGCGTCTGCAGAGGACAGAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 146735 MIM: 601311 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGFBP6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IGFBP6 - insulin like growth factor binding protein 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SOAT2 - sterol O-acyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003578.3 | 1160 | Missense Mutation | CGT,TGT | R,C 7 | NP_003569.1 | |
XM_011538849.2 | 1160 | Missense Mutation | CGT,TGT | R,C 7 | XP_011537151.1 | |
XM_011538850.2 | 1160 | Missense Mutation | CGT,TGT | R,C 7 | XP_011537152.1 | |
XM_011538851.2 | 1160 | Missense Mutation | CGT,TGT | R,C 7 | XP_011537153.1 |